Skip to Content
Merck
Altizide European Pharmacopoeia (EP) Reference Standard

Altizide

Altizide European Pharmacopoeia (EP) Reference Standard
Synonym(s):
Althiazide, 6-Chloro-3,4-dihydro-3-([2-propenylthio]methyl)-2H-1,2,4-benzothiadiazine-7-sulfonamide 1,1-dioxide
Empirical Formula (Hill Notation):
C11H14ClN3O4S3
CAS Number:
Molecular Weight:
383.89

Products

Formula Weight
Manufacturer
Format
Application
API Family
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing