Skip to Content
Merck
Allyl acetate 99%

Allyl acetate

Allyl acetate 99%
Synonym(s):
Allyl acetate
Linear Formula:
CH3COOCH2CH=CH2
CAS Number:
Molecular Weight:
100.12

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-2 of 2
Compare
Product Number
Description
Pricing