Skip to Content
Merck
Adrenaline with impurity F European Pharmacopoeia (EP) Reference Standard

Adrenaline with impurity F

Adrenaline with impurity F European Pharmacopoeia (EP) Reference Standard
Synonym(s):
(−)-Epinephrine, (−)-Adrenalin, (R)-(−)-3,4-Dihydroxy-α-(methylaminomethyl)benzyl alcohol, L-Adrenaline, L-Epinephrine
Linear Formula:
(HO)2C6H3CH(OH)CH2NHCH3
CAS Number:
Molecular Weight:
183.20

Products

Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Format
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing