
Adenosine Impurity G

Synonym(s):
Inosine, (−)-Inosine, Hypoxanthine 9-β-D-ribofuranoside
Empirical Formula (Hill Notation):
C10H12N4O5
CAS Number:
Molecular Weight:
268.23
Products
Shipping
Brand
Formula Weight
Melting Point (°C)
Physical Form
Quality Segment
Application
Package Size
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing