Skip to Content
Merck
7-Methoxy-4-(trifluoromethyl)coumarin ≥99% (TLC)

7-Methoxy-4-(trifluoromethyl)coumarin

7-Methoxy-4-(trifluoromethyl)coumarin ≥99% (TLC)
Synonym(s):
Methyl 4-(trifluoromethyl)umbelliferyl ether
Empirical Formula (Hill Notation):
C11H7F3O3
CAS Number:
Molecular Weight:
244.17

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing