![7-epi-Paclitaxel certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/223/277/cdc5d9d1-a9d3-41c5-8e68-51781e329496/640/cdc5d9d1-a9d3-41c5-8e68-51781e329496.png)
7-epi-Paclitaxel
![7-epi-Paclitaxel certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland](/deepweb/assets/sigmaaldrich/product/structures/223/277/cdc5d9d1-a9d3-41c5-8e68-51781e329496/640/cdc5d9d1-a9d3-41c5-8e68-51781e329496.png)
Synonym(s):
7-Epipaclitaxel
Empirical Formula (Hill Notation):
C47H51NO14
CAS Number:
Molecular Weight:
853.91
Products
Brand
Formula Weight
Manufacturer
Physical Form
Product Line
Quality Segment
Application
API Family
Reference Material Type
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing