5,10,15,20-Tetrakis(4-trimethylammoniophenyl)porphyrin tetra(p-toluenesulfonate)
Synonym(s):
TTMAPP
Empirical Formula (Hill Notation):
C84H90N8O12S4
CAS Number:
Molecular Weight:
1531.92
Products
Brand
Formula Weight
Physical Form
Chemical Composition
Quality Segment
Markush Class
Markush Group
Reaction Type
Reagent Type
Core
Showing 1-1 of 1
Compare
Product Number
Description
Pricing