Skip to Content
Merck
4-Methoxyphenol analytical standard

4-Methoxyphenol

4-Methoxyphenol analytical standard
Synonym(s):
4-Hydroxyanisole, 4-MP, HQMME, Hydroquinone monomethyl ether, MEHQ, p-Guaiacol, 4-Methoxyphenol
Linear Formula:
CH3OC6H4OH
CAS Number:
Molecular Weight:
124.14
EC Number:

Products

Shipping
Product Category
Brand
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Product Line
Purity
Format
Quality Segment
Application
Markush Class
Markush Group
Reference Material Type
Special Grade
Technique
Showing 1-4 of 4
Compare
Product Number
Description
Pricing