
(3S)-Boc-β-Pro-OH

Synonym(s):
(S)-1-Boc-3-pyrrolidinecarboxylic acid, (S)-1-(tert-Butoxycarbonyl)-3-pyrrolidinecarboxylic acid, N-Boc-D-β-proline
Empirical Formula (Hill Notation):
C10H17NO4
CAS Number:
Molecular Weight:
215.25
Products
Product Category
Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing