Skip to Content
Merck
3-(Methylphosphinico)propionic acid PESTANAL®, analytical standard

3-(Methylphosphinico)propionic acid

3-(Methylphosphinico)propionic acid PESTANAL®, analytical standard
Empirical Formula (Hill Notation):
C4H9O4P
CAS Number:
Molecular Weight:
152.09
EC Number:

Products

Product Category
Brand
Formula Weight
Product Line
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing