Skip to Content
Merck
2-Isopropylphenol ≥98%, FG

2-Isopropylphenol

2-Isopropylphenol ≥98%, FG
Synonym(s):
2-Isopropylphenol
Linear Formula:
(CH3)2CHC6H4OH
CAS Number:
Molecular Weight:
136.19

Products

Shipping
Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing