Skip to Content
Merck
All Photos(1)

Key Documents

PRI424050210

Princeton BioMolecular Research, Inc.

5-(6-methoxy-5-methylpyridin-3-yl)thiophene-2-carbaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
233.29
MDL number:

Iupac Name

5-(6-methoxy-5-methylpyridin-3-yl)thiophene-2-carbaldehyde

Inchi Code

1S/C12H11NO2S/c1-8-5-9(6-13-12(8)15-2)11-4-3-10(7-14)16-11/h3-7H,1-2H3

InChI key

LTJLHOOOWDUOQE-UHFFFAOYSA-N

Purity

95%

Country of Origin

UA

Physical Form

Solid

Product Link

http://store.princetonbio.com/?product_id=PBMR052752

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library