Skip to Content
Merck
All Photos(1)

Key Documents

KEY343107509

Key Organics / BIONET

3-hydroxy-2,3-dihydro-1??-benzothiophene-1,1-dione

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
184.22
MDL number:

Iupac Name

2,3-dihydro-1-benzothiophen-3-ol 1,1-dioxide

Inchi Code

1S/C8H8O3S/c9-7-5-12(10,11)8-4-2-1-3-6(7)8/h1-4,7,9H,5H2

InChI key

RVSZMOUZQCRNKH-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Liquid,Low Melting Solid

Product Link

https://www.keyorganics.net/sku/NF-0702

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

MSDS

https://www.keyorganics.net/sds/NF-0702


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service