ATC359510274
Atlantic Research Chemicals Ltd
4-Chloro-6-methyl-2-(1H-pyrrol-1-yl)pyrimidine
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Iupac Name
4-chloro-6-methyl-2-(1H-pyrrol-1-yl)pyrimidine
Inchi Code
1S/C9H8ClN3/c1-7-6-8(10)12-9(11-7)13-4-2-3-5-13/h2-6H,1H3
InChI key
WZMTWZSRVXQOHT-UHFFFAOYSA-N
Purity
95
Country of Origin
GB
Product Link
https://atlantic-chemicals.com/product/?code=HA01277
MSDS
https://atlantic-chemicals.com/product/?code=HA01277
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.