Skip to Content
Merck
All Photos(1)

Key Documents

APO455837440

Apollo Scientific Ltd

Allyl 1,1,2,2-tetrafluoroethyl ether

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Molecular Weight:
158.1
MDL number:
Pricing and availability is not currently available. Contact Technical Service

mp

-5

density

1.2

Iupac Name

3-(1,1,2,2-tetrafluoroethoxy)-1-propene

Inchi Code

1S/C5H6F4O/c1-2-3-10-5(8,9)4(6)7/h2,4H,1,3H2

InChI key

DTDFWYZVBJYVPL-UHFFFAOYSA-N

storage temp.

Ambient Temperature

Purity

98%

Country of Origin

GB

Shipping Temperature

Ambient Temperature

Physical Form

Liquid

Product Link

https://store.apolloscientific.co.uk/product/allyl-1122-tetrafluoroethyl-ether

Synonym

2-Propenyl 2H-tetrafluoroethyl ether

Flash Point

76-78

Melting Point

-5

density

1.2

Pictograms

Signal Word

Danger

Hazard Statements

Precautionary statements

MSDS

https://store.apolloscientific.co.uk/product/allyl-1122-tetrafluoroethyl-ether


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service