DWK241701401
DURAN® reaction flask with ground joint
glass flask, flask capacity (25 mL), single neck, round bottom, neck joint: ST/NS 19/26
Synonym(s):
241701401
All Photos(1)
About This Item
material
glass flask
round bottom
Joint
joint: ST/NS neck19/26
feature
lid
packaging
case of 10 ea
manufacturer/tradename
DURAN® 241701401
flask capacity
(25 mL)
General description
Tight closure due to standard ground joint. The geometry permits very uniform heating. Closable with glass and plastic stopper
Legal Information
DURAN is a registered trademark of DWK Life Sciences