Skip to Content
Merck
All Photos(1)

Documents

901218

Sigma-Aldrich

VPhos Pd G4

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C47H64NO4PPdS
CAS Number:
Molecular Weight:
876.47
UNSPSC Code:
12352202

Assay

95%

form

powder or crystals

feature

generation 4

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
reaction type: Cross Couplings

functional group

phosphine

Application

Buchwald palladacycle precatalyst with new biaryl(dialkyl)phosphine ligand, VPhos, has an improved activity profile for the reductive coupling of non-aromatic heterocyclic alkyl bromides. Its combined use with a surfactant system of octanoic acid/sodium octanoate enables Negishi coupling and access to diverse C(sp3)–C(sp2) cross-coupled products.

It is a fourth-generation (G4) Buchwald precatalysts are similar to the third generation (G3) precatalysts except that the amino group on the biphenyl backbone is methylated. This modification helps to prevent the limitations of using the third generation precatalysts. It is air-, moisture-, and thermally stable and shows good solubility in common organic solvents.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service