All Photos(1)
About This Item
Linear Formula:
FC6H3(CH3)CN
CAS Number:
Molecular Weight:
135.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Assay
99%
mp
50-52 °C (lit.)
functional group
fluoro
nitrile
SMILES string
Cc1ccc(F)c(c1)C#N
InChI
1S/C8H6FN/c1-6-2-3-8(9)7(4-6)5-10/h2-4H,1H3
InChI key
CMAOLVNGLTWICC-UHFFFAOYSA-N
General description
2-Fluoro-5-methylbenzonitrile is a phenolic cyanide derivative. Its vibrational spectra has been investigated. Vibrational investigations suggest that it belongs to the point group Cs.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Oral
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
201.2 °F - closed cup
Flash Point(C)
94 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Molecular structure, vibrational spectroscopic studies and analysis of 2-fluoro-5-methylbenzonitrile.
Sundaraganesan N, et al.
Indian Journal of Pure and Applied Physics, 47(7), 481-481 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service