Skip to Content
Merck
All Photos(1)

Documents

277371

Sigma-Aldrich

1-(Trimethylsilyl)pyrrolidine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H17NSi
CAS Number:
Molecular Weight:
143.30
Beilstein:
1304636
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.433 (lit.)

bp

139-140 °C (lit.)

density

0.821 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)N1CCCC1

InChI

1S/C7H17NSi/c1-9(2,3)8-6-4-5-7-8/h4-7H2,1-3H3

InChI key

NQLVIKZJXFGUET-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-(Trimethylsilyl)pyrrolidine (N-trimethylsilyl pyrrolidine) has been used in:
  • ring opening reaction of L-serine β-lactone to yield corresponding 5-amino-L-alanine derivatives
  • the preparation of iminium triflate salts

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

39.2 °F - closed cup

Flash Point(C)

4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service