HomeSupel™ Carbon LC U/HPLC Columns Demo Request
Supel™ Carbon LC U/HPLC Columns Demo Request
Porous Graphitic Carbon LC Column provides excellent retention properties for the analysis of polar substances.
- Elevated Temperature Stability: Columns can be readily operated at temperatures up to 250 °C, thereby allowing faster and more efficient separations.
- pH Stability: Compatible with mobile phases in the pH range of 1 – 14 at any temperature without causing a decline in the column lifespan.
- Unique retention mechanism: The Polar Retention Effect on Graphite (PREG) mechanism allows for the retention of polar or charged compounds without hydrophilic interaction liquid chromatography (HILIC) conditions. The mechanism also allows the resolution of geometric isomers.
- Compatibility with Any Solvent: Any polar or non-polar solvent can be used for the resolution of an analyte of interest.
- Stability to pressures up to 700 bar
We're here to help you sign up for a free demo of the Merck Supel™ Carbon LC column.
Supel™ Carbon LC Column specification |
---|
The Supel™ Carbon LC column comes in an internal diameter of 2.1, 3.0, and 4.6 mm, with a guard cartridge and holder for each internal diameter.
Materials
Loading
Register for a Free Demo
If you fill out the application form below, we will help you apply for counseling and free demo.