Merck
  • The Heme-Based Oxygen Sensor Rhizobium etli FixL: Influence of Auxiliary Ligands on Heme Redox Potential and Implications on the Enzyme Activity.
The Heme-Based Oxygen Sensor Rhizobium etli FixL: Influence of Auxiliary Ligands on Heme Redox Potential and Implications on the Enzyme Activity.
Journal of inorganic biochemistry (2016-09-12)
Nathalie Honorio-Felício, Marta S P Carepo, Tércio de F Paulo, Luiz Gonzaga de França Lopes, Eduardo H S Sousa, Izaura C N Diógenes, Paul V Bernhardt
ABSTRACT
Conformational changes associated to sensing mechanisms of heme-based protein sensors are a key molecular event that seems to modulate not only the protein activity but also the potential of the Fe
Sigma-Aldrich
2,5-Dihydroxy-1,4-benzoquinone, 98%