Merck
  • Theoretical investigation of the binding energies of the iodide ion and xenon atom with decaborane.
Theoretical investigation of the binding energies of the iodide ion and xenon atom with decaborane.
The journal of physical chemistry. A (2006-11-10)
Ilias Sioutis, Russell M Pitzer
ABSTRACT
The interaction of decaborane (B(10)H(14)) with the I(-) ion and the (isoelectronic) Xe atom is investigated using a number of theoretical methods: MP2, CCSD(T), CCSD, spin-orbit CISD, and DFT using the B3LYP, B3PW91, PW91PW91, and PBE0 methods. All non-DFT and some DFT methods agree that B(10)H(14)I(-) is bound by charge-dipole electrostatic forces, charge- and dipole-induced-dipole forces, and dispersion forces, while B(10)H(14)Xe is bound by dipole-induced-dipole forces and dispersion forces. Counterpoise corrections are necessary to obtain reliable results. Relativistic effective core potentials were used for the I, Xe, and B atoms. Basis sets for use with these potentials are discussed as is the question of basis set balance in molecules. We find B(10)H(14)I(-) to be bound by 19.8 kcal/mol and B(10)H(14)Xe by 1.1 kcal/mol, indicating that the charge and polarizability of I(-) play the major role in the interaction energy.
Sigma-Aldrich
Decaborane(14), white