Merck
  • [Analysis of the volatile oils chemical constituents of roots of Actinidia deliciosa].
[Analysis of the volatile oils chemical constituents of roots of Actinidia deliciosa].
Zhong yao cai = Zhongyaocai = Journal of Chinese medicinal materials (2008-10-02)
Han-Shen Zhen, Jing Ge, Jie Liang, Qin Qiu, Zhi-Hua Song, Zhen-Guo Zhong, Wen-Hui Wang, Jian-Ping Jiang
ABSTRACT
To analyze the volatile oils chemical constituents of roots of Actinidia deliciosa. The volatile oils fraction of roots of Actinidia deliciosa. were extracted by water vapor distilling, and then the constituents were separated and identified, by GC-MS. 16 compounds were identified, accounting for 89.37% of all quantity. The principal volatile oils chemical constituents are Phenol, 2,4-bis(1,1-dimethylethl)-; 2-Propenoic acid, 3-(4-methox yphenyl)-, ethyl ester; 9-Octadecenoic acid (Z)-, methyl ester; Cyclotetrasiloxane, octamethyl-.
Sigma-Aldrich
Octamethylcyclotetrasiloxane, 98%
SKU
Pack Size
Availability
Price
Quantity
Supelco
Isoamyl 4-methoxycinnamate, analytical standard