
Isoamyl 4-methoxycinnamate

Synonym(s):
4-Methoxycinnamic acid 3-methylbutyl ester, 4-Methoxycinnamic acid isoamyl ester, Amiloxate, Isopentyl 4-methoxycinnamate, NSC 408332
Empirical Formula (Hill Notation):
C15H20O3
CAS Number:
Molecular Weight:
248.32
Products
Brand
Formula Weight
Purity
Format
Quality Segment
Application
Reference Material Type
Special Grade
Technique
Showing 1-1 of 1