
4-Nitrophenyl N-acetyl-β-D-galactosaminide

Synonym(s):
4-Nitrophenyl 2-acetamido-2-deoxy-β-D-galactopyranoside, p-Nitrophenyl N-acetyl-β-D-galactosaminide
Empirical Formula (Hill Notation):
C14H18N2O8
Products
Product Category
Brand
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing