SAB5202482
Monoclonal Anti-4-Hydroxynonenal-Allophycocyanin antibody produced in mouse
clone 12F7
About This Item
Recommended Products
biological source
mouse
conjugate
Allophycocyanin conjugate
antibody form
purified immunoglobulin
antibody product type
primary antibodies
clone
12F7, monoclonal
form
buffered aqueous solution
species reactivity
human
concentration
1.0 mg/mL
technique(s)
immunofluorescence: 1:50
western blot: 1:1000
isotype
IgG1
shipped in
wet ice
storage temp.
−20°C
target post-translational modification
hydroxylation (4-Hydroxynonenal)
Immunogen
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
WGK 2
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service