Skip to Content
Merck
All Photos(2)

Key Documents

M8752

Sigma-Aldrich

6-Methyl-DL-tryptophan

>98% (TLC)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H14N2O2
CAS Number:
Molecular Weight:
218.25
Beilstein:
17684
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

product name

6-Methyl-DL-tryptophan, crystalline

Assay

>98% (TLC)

Quality Level

form

crystalline

color

light yellow to orange

mp

290 °C

storage temp.

2-8°C

SMILES string

Cc1ccc2c(CC(N)C(O)=O)c[nH]c2c1

InChI

1S/C12H14N2O2/c1-7-2-3-9-8(5-10(13)12(15)16)6-14-11(9)4-7/h2-4,6,10,14H,5,13H2,1H3,(H,15,16)

InChI key

GDMRVYIFGPMUCG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

6-Methyl-DL-tryptophan is a tryptophan analog.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Inge A Unsöld et al.
Microbiology (Reading, England), 151(Pt 5), 1499-1505 (2005-05-05)
A putative dimethylallyltryptophan synthase gene, fgaPT2, was identified in the genome sequence of Aspergillus fumigatus. fgaPT2 was cloned and overexpressed in Saccharomyces cerevisiae. The protein FgaPT2 was purified to near homogeneity and characterized biochemically. This enzyme was found to convert
Huixi Zou et al.
Journal of natural products, 72(1), 44-52 (2008-12-31)
This study reports that a series of tryptophan derivatives with modifications on the side chain or at the indole ring were accepted by two cyclic dipeptide prenyltransferases, CdpNPT and FtmPT1, and converted to prenylated derivatives. The structures of the enzymatic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service