EP6001040211
Eppendorf® Chest Freezer Rack
Holds 14 x 53 mm/2 in boxes, (14 x 1) configuration, stainless steel, side access, with locking rod
About This Item
Recommended Products
material
stainless steel
feature
(14 x 1) configuration
Holds 14 x 53 mm/2 in boxes
max. box size [mm]: 136 x 136
side access
with locking rod
manufacturer/tradename
Eppendorf® 6001040211 (Global)
technique(s)
cryopreservation: suitable
D × W × H
148 mm (5.8 in.) × 141 mm (5.6 in.) × 776 mm (30.6 in.)
capacity
(max box number per rack: 14 (1 x 14))
Looking for similar products? Visit Product Comparison Guide
General description
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service