EP5368000100
Eppendorf® Exchangeable Thermoblock
Holds 4 x DualChips
All Photos(1)
About This Item
UNSPSC Code:
41102404
NACRES:
NC.21
Recommended Products
feature
Holds 4 x DualChips
packaging
pkg of 1 ea
manufacturer/tradename
Eppendorf® 5368000100
Related Categories
General description
Product Information
For use with ThermoStat plus (heating/ cooling).
The exchangeable thermoblock system for the Eppendorf ThermoMixer® comfort and ThermoStat plus ensures a precise “fit” for all common lab tubes: from simple temperature control of a chemical to complex enzyme reactions and hybridization experiments, and from MTPs to 50 mL conical tubes – the application possibilities are virtually endless!
Features
Heat insulated thermoblocks for ergonomic and safe operation (no direct contact with hot surfaces - you won′t burn your fingers)
For use with ThermoStat plus (heating/ cooling).
The exchangeable thermoblock system for the Eppendorf ThermoMixer® comfort and ThermoStat plus ensures a precise “fit” for all common lab tubes: from simple temperature control of a chemical to complex enzyme reactions and hybridization experiments, and from MTPs to 50 mL conical tubes – the application possibilities are virtually endless!
Features
- A wide range of exchangeable thermoblocks to cover tubes from 0.2 mL to 50 mL as well as plates and glass slides
Heat insulated thermoblocks for ergonomic and safe operation (no direct contact with hot surfaces - you won′t burn your fingers)
- Dry incubation technology eliminates contamination risk associated with classical waterbath incubation
- Simple replacement of thermoblocks
- Tubes are easy accessible
- Whisper quiet operation and optimal temperature transfer due to complete enclosure and custom fit of tubes in the exchangeable thermoblocks
Legal Information
Eppendorf is a registered trademark of Eppendorf AG
Thermomixer is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)