Skip to Content
Merck
All Photos(1)

Key Documents

GF348

Sigma-Aldrich

TWEAK,
Human Recombinant Animal Free

Synonym(s):

TNFSF12, MGC129581, DR3LG, TWEAK, APO3L

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
eCl@ss:
32160405
NACRES:
NA.77

impurities

<0.1 μg/mg protein endotoxin (<1 EU/μg)

Quality Level

UniProt accession no.

shipped in

dry ice

General description

Product Source: Protein is expressed in E.coli.
The protein is manufactured without the use of any animal products/reagents and is considered Animal-Free and can be used in protocols where possible transmisson of animal viruses and contamination must be avoided.
TWEAK belongs to the TNF family of ligands and signals through TWEAKR also known as TNFRSF12A. TWEAK is expressed in a variety of tissues, including the adult heart, pancreas, skeletal muscle, small intestine, spleen and peripheral blood lymphocytes. TWEAK has the ability to induce NFkB activation and chemokine secretion, and to exert an apoptotic activity in certain cells, such as HT-29 human adenocarcinoma cells when cultured in the presence of IFN-. TWEAK also promotes proliferation and migration of endothelial cells.
Recombinant human TWEAK is a soluble 17.0 kDa polypeptide (154 amino acid residues) comprising the TNF homologous region of TWEAK and is generated by proteolytic processing of the full length membrane anchored TWEAK protein.

Application

Research Category
Stem Cell Research
Research Sub Category
Growth Factors & Receptors

Quality

Assay #1: The ED50 as determined by the dose-dependent stimulation of IL-8 production by human PBMC is less than 10 ng/ml.
Assay #2: TWEAK weakly induces the death of HT29 cells when cultured in the presence of IFN-γ. The ED50 for this effect is between 30-45 ng/ml.

Physical form

Product is presented in 10 mM Sodium Phosphate, pH 7.5 + 80
mM L-Arginine and is filtered through a 0.2 micron filter before lyophilization.

Storage and Stability

Store at -20°C for up to 4 months from date of receipt Centrifuge the vial prior to opening.
Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein and store in working aliquots at -20°C.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service