Key Documents
Z569984
Sigma-Aldrich
Aldrich® sleeve stopper septa, fluoroelastomer FKM
bottom I.D. × O.D. 12.7 mm × 23.7 mm, black, fits 24/40 joints and 64, 80 oz and 1gal jugs
About This Item
UNSPSC Code:
41121800
NACRES:
NB.35
Pricing and availability is not currently available.
material
fluoroelastomer FKM septa
bottom I.D. × O.D.
12.7 mm × 23.7 mm
color
black
General description
These septa are made from fluoroelastomer FKM which is highly resistant to a broad range of solvents.
An elastomeric study done at Sigma-Aldrich compared the solvent compatibility (resistance) of FKM and EPDM rubber, Fig. 1. A Sealability Factor was determined for each elastomer. Septa materials with lower sealability factor values, such as the Fluoroelastomer FKM, correlate with better solvent compatibility.
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Choose from one of the most recent versions:
Certificates of Analysis (COA)