Key Documents
Z115231
Scienceware® piezoelectric lighter
safe, useful tool for igniting Bunsen burners, gas stoves, propane, and oxyacetylene torches
Synonym(s):
piezo ignitor, piezo lighter, piezoelectric ignitor
About This Item
UNSPSC Code:
41100000
NACRES:
NB.35
Pricing and availability is not currently available.
manufacturer/tradename
Bel-Art
size
1 in. × 1 1/2 in. × 8 in.
General description
A safe, useful tool for igniting Bunsen burners, gas stoves, propane, and oxyacetylene torches. Produces a burst of 6 to 8 electric sparks at 20 kV without batteries, fuels, or flints. Plastic handle and a mounting bracket for convenient storage or hanging.
Other Notes
When using the product, the end of the barrel should be inserted in the middle of the actual gas flow, not off to the side. The gas needs to flow through the holes in the end of the barrel. Please note that the spark isn′t very large.
Legal Information
Scienceware is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)