Key Documents
SYNF41241L
Synthware two neck round bottom flask with angled side neck
1000 mL, center joint: ST/NS 24/40, side joint: ST/NS 24/40
Synonym(s):
Synthware F41241L, 2-neck round flask
About This Item
UNSPSC Code:
41121804
NACRES:
NB.17
Pricing and availability is not currently available.
material
borosilicate glass flask
round bottom
two-neck (angled side neck)
Joint
center joint: ST/NS24/40
side joint: ST/NS24/40
packaging
pkg of 1 ea
manufacturer/tradename
Synthware F41241L
capacity
1,000 mL
1000 mL
1000 mL
General description
Round bottom with joints. Heavy wall design. All sizes are hand-blown from tubing to ensure uniform wall thickness.
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)