BR365538
BRAND® volumetric flask for oil content determination
capacity 100 mL
Synonym(s):
Flasks, SILBERBRAND, Volumetric flasks
All Photos(1)
About This Item
UNSPSC Code:
41121804
NACRES:
NB.43
Recommended Products
material
glass
Joint
joint ST/NS neck19/26
feature
accuracy class B
packaging
pack of 1 ea
manufacturer/tradename
BRAND 365538
capacity
100 mL
Related Categories
General description
Borosilicate glass 3.3. To DIN 51368. Interchangeable hollow glass stopper size 19/26. Calibrated to contain (TC, In). For determination of oil content in aqueous oil emulsions (e.g. drilling oil). EL = Error limits.
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)