Skip to Content
Merck
All Photos(1)

Documents

213438

Sigma-Aldrich

Potassium triethylborohydride solution

1.0 M in THF

Synonym(s):

Potassium hydrotriethylborate, Potassium triethylhydridoborate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
KB(C2H5)3H
CAS Number:
Molecular Weight:
138.10
MDL number:
UNSPSC Code:
12352000
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reagent type: reductant

concentration

1.0 M in THF

density

0.902 g/mL at 25 °C

SMILES string

[K+].[H][B-](CC)(CC)CC

InChI

1S/C6H16B.K/c1-4-7(5-2)6-3;/h7H,4-6H2,1-3H3;/q-1;+1

InChI key

MFRDRDKPANBWOI-UHFFFAOYSA-N

Application

  • Reducing agent for synthesis of PtNi intermetallic nanoparticle catalysts with electrocatalytic activity
  • Reducing agent for preparation of vanadium nanoparticles (~7 nm) stabilized on activated carbon

Reagent for:
  • Preparation of lithium, sodium, and potassium boron monohydride complexes with tetramethylethylenediamine and pentamethyldiethylenetriamine
  • Dehydrogenation reactions
  • Tandem nucleophilic addition/C-C bond cleaving fragmentation reactions with cyclic vinylogous acyl triflates
  • Preparation of coordinatively unsaturated pseudotetrahedral iron(II) precursors as precatalysts for the room-temperature hydrogenation of unsaturated hydrocarbons
  • Substitution reactions

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 3

Flash Point(F)

1.4 °F - closed cup

Flash Point(C)

-17 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service