64840-U
Thimble
cellulose, size 25 mm × 80 mm
Synonym(s):
CELLULOSE THIMBLE 25 X 80,PK25
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
material
cellulose
packaging
pkg of 25
size
25 mm × 80 mm
application(s)
food and beverages
Looking for similar products? Visit Product Comparison Guide
General description
High-purity cellulose, approximately 1.5mm wall thickness, or high-quality borosilicate glass, 40-60μm porous disk.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service