TMO2403-0500
Nalgene® phthalate-free wash bottle
FEP, bottle volume 500 mL, unlabelled, case of 4 ea
Synonym(s):
fluid diepenser, plastic wash bottle
All Photos(1)
About This Item
Recommended Products
material
FEP
packaging
case of 4 ea
manufacturer/tradename
Nalgene 2403-0500
bottle volume
500 mL
General description
Ensure contamination-free dispensing with Thermo Scientific Nalgene Wash Bottles made with PTFE FEP, useful for trace element work. These phthalate-free bottles offer excellent chemical resistance and are molded entirely of fluoropolymers. Designed to dispense through the top with a closure and stem molded in one piece for uniform leakproof† service. The precision-molded tip provides a straight, steady stream and ultimate control without sputtering.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)