Skip to Content
Merck
All Photos(1)

Key Documents

SML2913

Sigma-Aldrich

Dehydroxylcannabidiol

≥98% (HPLC)

Synonym(s):

2-[(1R,6R)-3-Methyl-6-(1-methylethenyl)-2-cyclohexen-1-yl]-5-pentylphenol, DH-CBD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H30O
CAS Number:
Molecular Weight:
298.46
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

oil

optical activity

[α]/D -135 to -120°, c = 0.5 in chloroform-d

color

colorless to light brown

storage temp.

2-8°C

Biochem/physiol Actions

Dehydroxylcannabidiol (DH-CBD) is a synthetic nonpsychoactive cannabinoid that alleviate acute pain and chronic inflammatory pain by targeting α3 GlyRs. Dehydroxylcannabidio restores the function of both hyperekplexic mutant GlyRα1 and GABAAR in the presence of GlyRα1R271Q. Apparently dehydroxylcannabidiol interrupts the protein interaction GlyRα1 and GABAAR.
Synthetic nonpsychoactive cannabinoid that alleviate acute pain and chronic inflammatory pain by targeting α3 GlyRs

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wei Xiong et al.
The Journal of experimental medicine, 209(6), 1121-1134 (2012-05-16)
Certain types of nonpsychoactive cannabinoids can potentiate glycine receptors (GlyRs), an important target for nociceptive regulation at the spinal level. However, little is known about the potential and mechanism of glycinergic cannabinoids for chronic pain treatment. We report that systemic
Jieping Lu et al.
Neuropharmacology, 133, 224-232 (2018-02-07)
Some cannabinoids have been shown to suppress chronic pain by targeting glycine receptors (GlyRs). Although cannabinoid potentiation of α3 GlyRs is thought to contribute to cannabinoid-induced analgesia, the role of cannabinoid potentiation of α1 GlyRs in cannabinoid suppression of chronic
Guichang Zou et al.
The Journal of biological chemistry, 295(1), 138-145 (2019-11-24)
The functions of the glycine receptor (GlyR) and GABAA receptor (GABAAR) are both impaired in hyperekplexia, a neurological disorder usually caused by GlyR mutations. Although emerging evidence indicates that cannabinoids can directly restore normal GlyR function, whether they affect GABAAR

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service