Skip to Content
Merck
All Photos(1)

Key Documents

N0915

Sigma-Aldrich

NSC43067

≥98% (HPLC)

Synonym(s):

1-(5-Methyl-2-thienyl)-3-phenyl-2-Propen-1-one, 1-(5-Methyl-thiophen-2-yl)-3-phenyl-propenone, NSC 43067

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H12OS
CAS Number:
Molecular Weight:
228.31
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

light yellow to light tan

solubility

DMSO: >20 mg/mL

storage temp.

2-8°C

SMILES string

Cc1ccc(s1)C(=O)\C=C\c2ccccc2

InChI

1S/C14H12OS/c1-11-7-10-14(16-11)13(15)9-8-12-5-3-2-4-6-12/h2-10H,1H3/b9-8+

InChI key

AOUMOFQEBZGSPD-CMDGGOBGSA-N

Biochem/physiol Actions

NSC43067 is a Cyclin-dependent kinase (Cdk) inhibitor which binds to a novel conserved site on Cdk, presumably the p27 binding pocket. The binding apparently directly induces Cdk1, Cdk2, and Cdk4 protein aggregation. Cdks 1, 2, 4, and 6 are the key Cdks that regulate the cell cycle, and are a target for a cancer treatment. NSC43067 arrests the cell cycle at the G0/G1 phase at lower concentrations and at the G2/M cell cycle phase at higher concentrations. The action appears to be reversible.

Features and Benefits

This compound is featured on the CDKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diksha Kaushik et al.
International journal of pharmaceutics, 386(1-2), 42-51 (2009-11-11)
The enhancement/retardation of percutaneous permeation of diethyl-m-toluamide (DEET) in the presence of five percutaneous penetration modifiers (laurocapram, 3-dodecanoyloxazolidin-2-one (N-0915), S,S-dimethyl-N-(4-bromobenzoyl) iminosulfurane (DMBIS), S,S-dimethyl-N-(2-methoxycarbonylbenzenesulfonyl) iminosulfurane (DMMCBI) and tert-butyl 1-dodecyl-2-oxoazepan-3-yl-carbamate (TBDOC)) was investigated. These permeation modifiers were formulated in either water, propylene

Articles

Review properties, activators and inhibitors, and available products for researching cyclin-dependent kinases (CDKs).

Review properties, activators and inhibitors, and available products for researching cyclin-dependent kinases (CDKs).

Review properties, activators and inhibitors, and available products for researching cyclin-dependent kinases (CDKs).

Review properties, activators and inhibitors, and available products for researching cyclin-dependent kinases (CDKs).

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service