Skip to Content
Merck
All Photos(1)

Key Documents

M4438

Sigma-Aldrich

Anti-Munc-18 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 67 kDa

species reactivity

rat

technique(s)

immunoprecipitation (IP): suitable
western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

Immunogen

synthetic peptide corresponding to amino acids 58-70 of human Munc-18.

Physical form

Solution in phosphate buffered saline, containing 1 mg/ml bovine serum albumin and 0.05% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bianca Karine Marin et al.
Experimental physiology, 104(7), 1029-1037 (2019-04-27)
What is the central question of this study? Does protein restriction in early life modify glucose-induced insulin secretion by altering [Ca2+ ]i and the expression of SNARE proteins in pancreatic islets from pregnant rats? What is the main finding and
Carla Bruna Pietrobon et al.
The Journal of physiology, 598(3), 489-502 (2019-12-13)
The World Health Organization recommends exclusive breastfeeding until 6 months of age as an important strategy to reduce child morbidity and mortality. Studies have associated early weaning with the development of obesity and type 2 diabetes in adulthood. In our model

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service