Skip to Content
Merck
All Photos(1)

Key Documents

K3389

Sigma-Aldrich

Anti-Ku80 (XRCC5) (322-337) antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-ATP-dependent DNA Helicase II, 80kDa, Anti-CTCBF (CTC85), Anti-Ku Autoantigen, 80kDa, Anti-Lupus Ku Autoantigen Protein p86, Anti-X-ray Repair Complementing Defective Repair in Chinese Hamster Cells 5

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~82.5 kDa by calculation

species reactivity

human

technique(s)

western blot: 1:500

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... XRCC5(7520)
mouse ... Xrcc5(22596)
rat ... Xrcc5(363247)

Immunogen

peptide corresponding to the 80 kDa Ku80 protein (amino acids 322-337). This sequence is 100% conserved between human, mouse, rat, and hamster.

Physical form

Solution of 1 mg/mL in phosphate buffered saline containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Loraine Jarrosson et al.
iScience, 24(12), 103423-103423 (2021-12-02)
Lack of preclinical patient-derived xenograft cancer models in which to conduct large-scale molecular studies seriously impairs the development of effective personalized therapies. We report here an in vivo concept consisting of implanting human tumor cells in targeted tissues of an avian

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service