Skip to Content
Merck
All Photos(1)

Key Documents

EHU009241

Sigma-Aldrich

MISSION® esiRNA

targeting human ITGB6 (2)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105324
NACRES:
NA.51

description

Powered by Eupheria Biotech

Quality Level

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CAAACTAGCAGGCATCGTCATTCCTAATGACGGGCTCTGTCACTTGGACAGCAAGAATGAATACTCCATGTCAACTGTCTTGGAATATCCAACAATTGGACAACTCATTGATAAACTGGTACAAAACAACGTGTTATTGATCTTCGCTGTAACCCAAGAACAAGTTCATTTATATGAGAATTACGCAAAACTTATTCCTGGAGCTACAGTAGGTCTACTTCAGAAGGACTCCGGAAACATTCTCCAGCTGATCATCTCAGCTTATGAAGAACTGCGGTCTGAGGTGGAACTGGAAGTATTAGGAGACACTGAAGGACTCAACTTGTCATTTACAGCCATCTGTAACAACGGTACCCTCTTCCAACACCAAAAGAAATGCTCTCACATGAAAGTGGGAGACACAGCTTCCTTCAGCGTGACTGTGAATATCCCACACTGCGAGAGAAGAAGCAGGCACATTATCATAAAGCCTGTGGGGCTGGGGGATGCCCTGGAATTACTTGTCAGCCCAGAATGCAACTGCGACTGTCAG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert J Slack et al.
Pharmacology, 97(3-4), 114-125 (2016-01-07)
A20FMDV2 is a peptide derived from the foot-and-mouth disease virus with a high affinity and selectivity for the alpha-v beta-6 (αvβ6) arginyl-glycinyl-aspartic acid (RGD)-binding integrin. It has been shown to be an informative tool ligand in pre-clinical imaging studies for
Jiarui Bi et al.
Cytokine, 114, 135-142 (2018-11-24)
Epithelial αvβ6 integrin participates in immune surveillance in many organs, including the gastrointestinal track. Expression of αvβ6 integrin is reduced in the junctional epithelium of the gingiva in periodontal diseases, and mutations in the ITGB6 gene are associated with these
Jiarui Bi et al.
Scientific reports, 7(1), 4411-4411 (2017-07-02)
Periodontal diseases manifest by the formation of deep pockets between the gingiva and teeth where multispecies bacterial biofilms flourish, causing inflammation and bone loss. Epithelial cell receptor αvβ6 integrin that regulates inflammation by activating the anti-inflammatory cytokine transforming growth factor-β1
Runhong Han et al.
JCI insight, 4(7) (2019-04-05)
Chronic tubulointerstitial injury impacts the prognosis of focal segmental glomerulosclerosis (FSGS). We found that the level of versican V1 was increased in tubular cells of FSGS patients. Tubular cell-derived versican V1 induced proliferation and collagen synthesis by activating the CD44/Smad3

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service