Recommended Products
grade
certified reference material
Quality Level
packaging
pkg of 100 g
manufacturer/tradename
NIST®
format
matrix material
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
SRM 634A_cert
SRM 634A_SDS
Application
Features and Benefits
- NIST certified values for elements in cement are provided and the values are metrologically traceable to SI unit of mass, expressed as a percentage.
- Reference values and information values for constituents in SRM634a are reported as mass fraction on an received basis.
- Accompanied with maintenance and expiration of SRM certification and instructions for use.
Other Notes
Silicon dioxide (SiO2), Aluminium oxide (Al2O3), Ferric oxide (Fe2O3), Calcium oxide (CaO), Magnesium oxide (MgO), Sulphur Trioxide (SO3), Potassium oxide (K2O), Titanium dioxide (TiO2), Phosphorus pentoxide (P2O5)
Reference/Informational value is provided for the following analytes.
Sodium oxide (Na2O), Chromium(III) oxide (Cr2O3), Zinc oxide (ZnO), Manganese(III) oxide (Mn2O3), Strontium oxide (SrO), Free Calcium oxide (CaO)
See certificate for values and more details at nist.gov/SRM.
Legal Information
related product
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
13 - Non Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service