Skip to Content
Merck
All Photos(1)

Key Documents

ERMCE100

Fish tissue (HCB and HCBD)

ERM®, certified reference material

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material

Quality Level

Agency

ERM®

manufacturer/tradename

JRC

application(s)

food and beverages

format

matrix material

storage temp.

2-8°C

General description

Hexachlorobenzene (HCB) and hexachlorobutadiene (HCBD)

Analysis Note

For more information please see:
ERMCE100

Legal Information

ERM is a registered trademark of European Commission

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service