Skip to Content
Merck
All Photos(3)

Key Documents

857408

Sigma-Aldrich

Acid Fuchsin calcium salt

certified by the Biological Stain Commission, Dye content ≥60 %

Synonym(s):

Acid Magenta, Acid Rubin, Fuchsin S, SS, SS, ST, or SIII, Rubin S

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C20H17N3O9S3Ca
CAS Number:
Molecular Weight:
579.64
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
NA.47

grade

certified by the Biological Stain Commission

Quality Level

form

powder or crystals

composition

Dye content, ≥60%

technique(s)

microbe id | staining: suitable

λmax

545 nm

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Ca++].Cc1cc(cc(c1N)S([O-])(=O)=O)\C(c2ccc(N)c(c2)S([O-])(=O)=O)=C3\C=CC(=N)C(=C3)S(O)(=O)=O

InChI

1S/C20H19N3O9S3.Ca/c1-10-6-13(9-18(20(10)23)35(30,31)32)19(11-2-4-14(21)16(7-11)33(24,25)26)12-3-5-15(22)17(8-12)34(27,28)29;/h2-9,21H,22-23H2,1H3,(H,24,25,26)(H,27,28,29)(H,30,31,32);/q;+2/p-2/b19-11-,21-14?;

InChI key

SVIISRNRODSEHG-SZRYGYPJSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Acid fuchsine (C.I. 42685) is a sulfonated triaminotriphenylmethane. It is also known as acid magenta. At pH 12-14, acid fuchsine changes from red to colorless.

Application

Acid fuchsine is used as a biological stain in preparations including the histopathological polychrome of Pianese, the Ehrlich-Biondi triacid procedure for blood smears, and the Altmann stain for mitochondria. Various polychrome stains like the Masson polychrome variant; Van Gieson, and Mallory connective tissue stains contain Acid fuchsine.Other applications of acid fuchsine in biology and medicine involve fluorescent staining of latent fingerprints, the tracking of water flow within plants, and adding into an agar growth medium for yeasts.

Suitability

Certified for use in Van Gieson connective tissue stain.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Analysis and testing of biological stains--the Biological Stain Commission Procedures.
D P Penney et al.
Biotechnic & histochemistry : official publication of the Biological Stain Commission, 77(5-6), 237-275 (2003-02-05)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service