Skip to Content
Merck
All Photos(1)

Key Documents

870723P

Avanti

23:0 Coenzyme A

Avanti Research - A Croda Brand 870723P, powder

Synonym(s):

tricosanoyl Coenzyme A (ammonium salt)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H89N10O17P3S
CAS Number:
Molecular Weight:
1155.22
MDL number:
UNSPSC Code:
12352211
NACRES:
NA.25

form

powder

packaging

pkg of 1 × 5 mg (870723P-5mg)

manufacturer/tradename

Avanti Research - A Croda Brand 870723P

application(s)

lipidomics

lipid type

coenzymes

shipped in

dry ice

storage temp.

−20°C

SMILES string

O[C@@](C(NCCC(NCCSC(CCCCCCCCCCCCCCCCCCCCCC)=O)=O)=O)(C(C)(COP([O-])(OP([O-])(OC[C@H]([C@H]1OP([O-])(O)=O)O[C@H]([C@@H]1O)N2C3=C(C(N)=NC=N3)N=C2)=O)=O)C)[H].[NH4+].[NH4+].[NH4+]

InChI

1S/C44H80N7O17P3S.3H3N/c1-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-19-20-21-22-23-24-35(53)72-28-27-46-34(52)25-26-47-42(56)39(55)44(2,3)30-65-71(62,63)68-70(60,61)64-29-33-38(67-69(57,58)59)37(54)43(66-33)51-32-50-36-40(45)48-31-49-41(36)51;;;/h31-33,37-39,43,54-55H,4-30H2,1-3H3,(H,46,52)(H,47,56)(H,60,61)(H,62,63)(H2,45,48,49)(H2,57,58,59);3*1H3/t33-,37?,38+,39+,43-;;;/m1.../s1

InChI key

VWUVJQXOYHTDCP-BHMMHRQOSA-N

General description

23:0 Coenzyme A, also known as tricosanoyl coenzyme A, is a coenzyme A derivative of tricosylic acid.

Application

23:0 Coenzyme A has been used as a substrate in acylglycerol-3-phosphate-O-acyltransferases (AGPAT 11) assay and ceramide synthase assay.

Packaging

5 mL Amber Glass Screw Cap Vial (870723P-5mg)

Legal Information

Avanti Research is a trademark of Avanti Polar Lipids, LLC

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anil K Agarwal et al.
Journal of lipid research, 51(8), 2143-2152 (2010-04-07)
The conversion of lysophosphatidic acid (LPA) to phosphatidic acid is carried out by the microsomal enzymes 1-acylglycerol-3-phosphate-O-acyltransferases (AGPATs). These enzymes are specific for acylating LPA at the sn-2 (carbon 2) position on the glycerol backbone and are important, because they
Evgeny V Berdyshev et al.
The Journal of biological chemistry, 284(9), 5467-5477 (2009-01-03)
Novel immunomodulatory molecule FTY720 is a synthetic analog of myriocin, but unlike myriocin FTY720 does not inhibit serine palmitoyltransferase. Although many of the effects of FTY720 are ascribed to its phosphorylation and subsequent sphingosine 1-phosphate (S1P)-like action through S1P(1,3-5) receptors

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service