Z232637
Duran® sintered disc filter funnel
capacity 1,000 mL, nominal max pore size 10-16 μm
Synonym(s):
258524403
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104900
NACRES:
NB.43
Recommended Products
material
(Both funnels and frits are made from borosilicate glass)
manufacturer/tradename
DURAN®
Schott
availability
not available in USA and Canada
capacity
1,000 mL
disc diam.
120 mm
filter diam.
120 mm
pore size
10-16 μm nominal max pore size
Looking for similar products? Visit Product Comparison Guide
General description
Especially useful where filter paper would be attacked by filtered material.
Sintered discs with max. pore sizes in the range 40 to 100 μm are often used for preparative work and mercury filtration, 16 to 40 μm discs for analytical work with medium precipitates and 10 to 16 μm for fine precipitates. 1.0 to 1.6 μm discs are for ultra-fine filtration of biological solutions as they remove bacteria from liquids that would decompose if heated.
Porosity chart for Duran filter funnels:
Grade 2: 40-100 μm nominal max. pore size
Grade 3: 16-40 μm nominal max. pore size
Grade 4: 10-16 μm nominal max. pore size
Grade 5: 1.0-1.6 μm nominal max. pore size
Sintered discs with max. pore sizes in the range 40 to 100 μm are often used for preparative work and mercury filtration, 16 to 40 μm discs for analytical work with medium precipitates and 10 to 16 μm for fine precipitates. 1.0 to 1.6 μm discs are for ultra-fine filtration of biological solutions as they remove bacteria from liquids that would decompose if heated.
Porosity chart for Duran filter funnels:
Grade 2: 40-100 μm nominal max. pore size
Grade 3: 16-40 μm nominal max. pore size
Grade 4: 10-16 μm nominal max. pore size
Grade 5: 1.0-1.6 μm nominal max. pore size
Features and Benefits
- Manufactured from 3.3. expansion borosilicate glass
- Funnels have an integral sintered glass filtration disc
- Comprehensive range of funnel sizes and sinter porosities (grades) are available
- The use of filter paper is not required
- Resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
- Filter funnels can be used with a filtering flask via a conical rubber seal (GUKO)
- Fired-on, highly durable white print
Legal Information
DURAN is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service