Skip to Content
Merck
All Photos(1)

Key Documents

SLW4513/60M

Pyrex® Culture tube cap inserts

thread size 60 mm

Synonym(s):

Pyrex, SciLabware 4513/60M

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121703
NACRES:
NB.11

thread size

60 mm

manufacturer/tradename

4513/60M
SciLabware

Looking for similar products? Visit Product Comparison Guide

Features and Benefits

• PBT cap with PTFE faced cap liner• Withstands hot air sterilisation up to 180 °C• Autoclavable• Cap liner is ethylene propylene with PTFE face for chemical resistance.

Other Notes

Insert Only

Legal Information

PYREX is a registered trademark of Corning, Inc.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service