SLW3750/03
Pyrex® Filter funnel with sintered glass disc
30 mL, bowl I.D. 80 mm, porosity grade 3
Synonym(s):
Pyrex, SciLabware 3750/03
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104900
NACRES:
NB.43
Recommended Products
material
borosilicate glass
manufacturer/tradename
3750/03
SciLabware
bowl I.D.
80 mm
capacity
30 mL
30 mL
stem O.D. × L
8 mm × 82 mm
pore size
grade 3 porosity
Looking for similar products? Visit Product Comparison Guide
General description
- Sintered disc manufactured from Pyrex borosilicate glass
- Available in a choice of porosities
For the filtration of solutions:
- No requirement for filter paper
- Resistant to most corrosive reagents including ammonia and concentrated sulphuric acid
Legal Information
PYREX is a registered trademark of Corning, Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service