DWKWGW233P
Azlon Square Shoulder Unvented Wash Bottles
wide-neck, low-density polyethylene bottle, green polypropylene closure, capacity 250 mL
Synonym(s):
WGW233P
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
green polypropylene closure
low-density polyethylene bottle
wide-neck
packaging
pkg of 5 ea
manufacturer/tradename
Azlon WGW233P
capacity
250 mL
Looking for similar products? Visit Product Comparison Guide
Application
Ideal for solvent and water dispensing within the laboratory
Features and Benefits
Precision jet tip for fine control
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service